missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and ion content of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
- Large color touchscreen provides direct measurement and easy data entry.
- Instructions guide users through the sensor calibration process.
- View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- A two-level user management protects the most important settings.
- The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
- Dedicated positions for cleaning and storage support correct sensor handling.
Specifications
Specifications
| Accuracy (Ion) | 0.5 % |
| Accuracy (mV) | 0.1 mV |
| Accuracy (pH) | 0.002 |
| Data Management Details | PC, Printer, USB Stick |
| Description | SevenDirect SD50 Kit |
| Channels | 1 |
| Display Type | Color TFT |
| IP Rating | IP54 |
| Measurement Modes | pH |
| Range (mV) | -2000 to 2000 mV |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
