missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ pH Meter
Description
With case, batteries, buffers and screw driver 1.00pH to 13.00pH 90mm electrode length Hanna
Specifications
Specifications
| Accuracy (pH) | ±0.01 |
| Range (pH) | 1 to 13 |
| Temperature Range (Metric) | 0°C to 70°C |
| Resolution | ±0.01 |
| Type | pH Stick Piccolo |
| Width (Metric) | 190 mm |
| Height (Metric) | 15 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?