missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ pH Paper

Product Code. 10797941 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
0.5 to 5.5
1 to 11
1 to 14
3.8 to 5.8
4 to 7
5.4 to 7
5.5 to 9
6.4 to 8
7.2 to 9.7
9 to 13
Unit Size:
5m
Each
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10797941 1 to 14 5m
10098250 0.5 to 5.5 Each
10464491 1 to 11 Each
10323361 3.8 to 5.8 5m
10539641 4 to 7 Pack of 5
10549641 5.4 to 7 Pack of 5
10559641 5.5 to 9 5m
10570022 6.4 to 8 5m
10613141 7.2 to 9.7 5m
10527341 9 to 13 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10797941 Supplier Macherey-Nagel™ Supplier No. 90204
  • ONLINE EXCLUSIVE PRICE
    13.20 €
    11.51 € / 5m
    Save 1.69 €
    12% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.

 
  • Universal and special indicator papers - simple pH measurement
  • Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
  • Always ready for use .
  • The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .

Specifications

Certifications/Compliance ISO 9001
pH Range 1 to 14
Format Roll
Graduations 1, 2, 3, 5, 6, 7, 8, 9, 10, 12, 14
Quantity 1 Each
Type pH Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.