missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ pH and pH/ORP Sensors for Model 556 Multi-Probe System
Click to view available options
Type:
pH Probe
pH and pH/ORP probe
Unit Size:
Each
Description
- User-replaceble sensor allows for quick sensor replacements if needed
- No tools necessary
Specifications
Specifications
| Type | pH and pH/ORP probe |
| For Use With (Application) | Aquaculture, agriculture, chemistry, food and beverage, wine-making and water-testing, for scientific applications |
| For Use With (Equipment) | YSI model 556 handheld multi probe meter |
| Includes | Storage bottle with storage solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction