Learn More
Testo™ pH/Temperature Meter 206-pH1 Starter Kit
Everything needed to start measuring the temperature and pH value in liquids such as drinking or heating water.
Brand: Testo™ 0563 2065
Code : 04
Additional Details : Weight : 2.50000kg Transport : UN number : 3091 Chem class : 9
Description
- Ideal for measuring the pH value of liquids
- Temperature measurement is feasible thanks to the integrated temperature probe
- Robust and waterproof thanks to TopSafe™ protective case with IP68
- Set contains essential accessories such as calibration dosing bottles, aluminum case, storage cap, etc.
- Precise, fast temperature compensation
- Maintenance-free, leak-proof and insensitive thanks to gel electrolyte
- Dual wall diaphragm of pH probe
- Simultaneous display of pH and temperature measurements
Specifications
2 measurements per second | |
±0.4°C | |
0 to 14 | |
0°C to 60°C (Short-term to 80°C Max. 5 min.) | |
LCD | |
197 x 33 x 20 mm | |
ABS Plastic | |
2 | |
80 hr. (Auto Off after 10 min.) |
±0.02 | |
Storage cap with gel, 2 calibration bottles each 250mL, protective case of TopSafe™, belt/wall bracket, aluminum case | |
0.01 | |
0.1°C | |
IP-68 | |
69 g | |
0°C to 60°C | |
1 x CR2032 |