missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Derm ASEPT Hand Soap

Product Code. 17830265
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
Quantity:
12 Airless Bottles
20 Square Mounted Pump Bottles
25 Mounted Pump Bottles
Unit Size:
10 L
12 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Quantity Unit Size
17830265 1000 mL 12 Airless Bottles 12 L
17870245 500 mL 20 Square Mounted Pump Bottles 10 L
17880245 1000 mL 25 Mounted Pump Bottles 12 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17830265 Supplier Christeyns™ Supplier No. C65122
  • ONLINE EXCLUSIVE PRICE
    186.00 €
    166.62 € / 12 L
    Save 19.38 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phago'Derm ASEPT is a disinfectant cleansing foam soap, specially formulated for hygienic hand washing before any aseptic procedure (insertion of catheters, etc.) and systematically before and after any contact with a patient or contaminating material.

  • Formula keeps epidermis soft, even in the event of frequent washing
  • Enriched with glycerin
  • Specially formulated for the medical, food and institutional sectors

Specifications

Product Type Anti-bacterial Hand Soap
Color Light Beige
Form Foam
Quantity 12 Airless Bottles
Volume (Metric) 1000 mL
compliance-icons compliance-icons
Product Identifier
  • PHAGO'DERM ASEPT
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.