missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Soft Disinfectant Detergent

Product Code. 17831135
Change view
Click to view available options
Product Type:
Disinfectant Solution
Disinfectant Surface Spray
Size:
750 mL
5 L
Unit Size:
10 L
4.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Size Unit Size
17831135 Disinfectant Solution 5 L 10 L
17821135 Disinfectant Surface Spray 750 mL 4.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17831135 Supplier Christeyns™ Supplier No. C65006
  • ONLINE EXCLUSIVE PRICE
    66.90 €
    60.20 € / 10 L
    Save 6.70 €
    10% Off
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phago'Soft is a disinfectant detergent for all high surfaces, glass surfaces and screens. It is also compatible with stainless steel.

  • Ready to use
  • Eco-certified
  • Approved for food contact
  • Active substance (peracetic acid) guarantees broad-spectrum microbiological effectiveness while minimizing its environmental impact through its degradation into acetic acid, oxygen and water

Specifications

Product Type Disinfectant Solution
Color-coded No
Form Clear Liquid
pH 3 to 3.7
Scent Fresh Scent
Sterility Non-sterile
Quantity 2 Cans
Size 5 L
compliance-icons
Product Identifier
  • 10LT PHAGOSOFT 5LT EcoCert (2X5LT)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, hydrogen peroxide, acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.