missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Spray DM

Product Code. 16612734
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
16612734 5 L Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16612734 Supplier Christeyns™ Supplier No. 60414
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Phago'Spray DM disinfectant boasts broad spectrum efficacy on rotavirus in 5 minutes.

  • Professional disinfectant cleaner for the disinfection of surfaces in high-risk environments
  • Can be used as a spray on walls, ceilings and furniture in care and surgery departments
  • Can also be used in application on a single-use cloth for the rapid disinfection of small furniture, trolleys, exterior surfaces of non-invasive medical devices and small surfaces close to the patient
  • Ready-to-use
  • Includes nozzle with two positions (spray and stop pictograms)
  • For use in foam or spray

Specifications

Product Type Disinfectant Cleaner
Color-coded No
Form Clear Liquid
pH Neutral
Scent Original
Sterility Non-sterile
Quantity 2 Cans
Size 5 L
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • phago'spray DM
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, didecyldimethylammonium chloride, N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, ethane-1,
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.