missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Phalloidin-TRITC

Click to view available options
Quantity:
12 μg
Unit Size:
12µg
Description
Phalloidin-TRITC is a red-orange fluorescent cytoskeleton stain. Binds and labels F-actin. For most fluorescent imaging applications, cells should to be fixed and permeablilized. Excitation/emission maxima λ ∼ 540/565 nm
We suggest making up a stock solution of this product by dissolving the vial contents in 1.5 mL methanol or DMSO. Please refer to the protocol for further information on using this product.
Specifications
Specifications
| Chemical Name or Material | 7-[(4R)-5-[[[[4-[3,6-Bis(dimethylamino)xanthylium-9-yl]-3-carboxyphenyl]amino]thioxomethyl]amino]-4-hydroxy-L-leucine]phalloidin |
| CAS | 915013-10-4 |
| Quantity | 12 μg |
| Target | Fluorescent Actin Probes |
| Molecular Formula | C60H70N12O13S2 |
| Formula Weight | 1231.4 |
| Purity | 90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction