missing translation for 'onlineSavingsMsg'
Learn More

Pharma MagMax™ Express-96 Tip Combs for PCR Head

Product Code. 31759208 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
80 combs
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31759208 80 combs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31759208 Supplier ABI Applied Biosystems Supplier No. 4472784
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These are replacement tip combs for the Pharmaceutical Analytics KingFisher™ Flex 96 PCR Head Magnetic Particle Processor and MagMAX™ Express-96 Deep Well Magnetic Particle Processor with PCR Head.

These are replacement tip combs for the Pharmaceutical Analytics KingFisher™ Flex 96 PCR Head Magnetic Particle Processor and MagMAX™ Express-96 Deep Well Magnetic Particle Processor with PCR Head.

Optimize performance using MagMax/KingFisher system consumables designed specifically for use with these instruments. MagMax/KingFisher instruments utilize disposable plastics made of polypropylene. The plastics—tip combs and microplates—are ideal for magnetic particle processing because of their low binding affinity for biomolecules. The special design enables excellent recovery of magnetic beads.

Contains eighty (80) pieces dual-packaged in ten (10) paperboard boxes packed into one (1) cardboard box.

Specifications

Format 96-well Plate
Capacity 96 Samples
Content And Storage Contains eighty (80) pieces dual-packaged in ten (10) paperboard boxes packed into one (1) cardboard box.
Product Line MagMAX
Quantity 80 combs
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.