Learn More
Pharma Magnetic Stand-96
Product Code. 30485110 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Stand
Unit Size:
Each
Product Code. Quantity Unit Size
30485110 1 Stand Each
1 options
Product Code. 30485110 Supplier ABI Applied Biosystems Supplier No. A31543
  • ONLINE EXCLUSIVE PRICE
    889.00 €
    858.00 € / Pack of 1
    Save 31.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Magnetic Stand-96 is designed for paramagnetic bead precipitation from standard 96-well, U-bottom microplates and 0.2 mL PCR plates with no additional accessories.
The stand has a clear polyacryl base in a standard SBS 96-well microplate footprint. The use of 24 extremely strong permanent magnets enables easy and fast magnetic separation in as little as 30 seconds. It sits flat on a lab bench as well as in a microplate adapter on robotic workstations.
TRUSTED_SUSTAINABILITY
Specifications
Shipping Condition Room Temperature
Product Line KingFisher™ Flex
Quantity 1 Stand
Type Magnetic Stand