Learn More
Pharmaceutical Analytics QuantStudio™ 5 Real-Time PCR System, 384-well, laptop
Shop All Applied Biosystems ProductsDescription
The Applied Biosystems™ QuantStudio™ 5 Real-Time PCR System for Pharmaceutical Analytics is designed for users who need superior performance, maximum dye versatility, and security options in a real-time PCR system that is affordable and easy to use. The QuantStudio 5 pharmaceutical analytics system is used together with specialized sample preparation and quantitation kits for host cell protein, host cell DNA, and Protein A impurity testing of biological products produced in cell culture.The optimized Design and Analysis software is ideal for both first-time and experienced users. When connected to the Thermo Fisher Cloud, the QuantStudio 5 system provides access to your data wherever and whenever you want. Using proven OptiFlex™ technology (featuring 5 channels and white LED), the QuantStudio 5 384-well system enables improved data accuracy and sensitivity for a broad range of genomic applications. The QuantStudio 5 system also offers the built-in software features of electronic record security and prevention of unauthorized instrument access to assist with 21 CFR Part 11 compliance. For validated application solutions, please see Cat. No. A31672.
Features of the QuantStudio 5 system with 384-well block include:
- Proven OptiFlex technology (5 channels/21 filter combinations with white LED) for trusted results
- User account management, locked workflow, and Pause features for greater control of experiment data
- Built-in software features to support compliance with 21 CFR Part 11 guidelines
- Secure cloud-based software provides increased accessibility and storage options
- Simplified instrument software with interactive touchscreen for greater ease of use
- 384-well format
Thermo Fisher Cloud
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Upload data from the instrument or upload your own files from anywhere you have web access. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use.
Easy to use
The QuantStudio 5 Real-Time PCR System is compact in form, factory calibrated and is delivered in user-friendly packaging to enable fast and simple installation for immediate use. The interactive touchscreen provides a modern usability experience and the simplified instrument software/pre-optimized protocol templates allow users of any experience level to quickly set up and start a run.
Design and analysis software options:
Option 1: QuantStudio Design and Analysis cloud software
- Supports web browser-based system configuration with PC or MAC computer
- Streamlined software for improved usability and response time
- Enables secure access of your data when and where you want through the cloud
- No software to install, no licenses and no versions to update
Option 2: QuantStudio Design and Analysis desktop software
- Supports traditional co-located computer system configuration on Windows™ 10 IOT OS
- Streamlined software for improved usability and response time
- No licenses to purchase
Total control
With built-in software features to support compliance with 21 CFR Part 11 guidelines, user account management, locked workflow and Pause features, the QuantStudio 5 system offers users greater control of experiment data.
Multiplex performance results that you can trust
Improved well-to-well and instrument-to-instrument data accuracy is achieved with OptiFlex technology. In the 384-well system this technology features 5 color excitation and emission filter channels, for a total of 21 filter combinations, with white LED.
Sensitive data analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
High application versatility
Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, and more.
Specifications
Specifications
| Block Format | Non-interchangeable |
| Format | 384-well Plate |
| Content And Storage | QuantStudio 5 Real Time PCR System, 384-well, Laptop |
| Display Type | Touchscreen |
| Shipping Condition | Room Temperature |
| Quantity | 1 system |
Frequently Asked Questions (FAQs)
We recommend trying different browsers (Google Chrome, Firefox, and Microsoft Edge) and/or removing cookies from your browser. If this does not resolve the issue, please contact Technical Support at techsupport@thermofisher.com.
If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.