missing translation for 'onlineSavingsMsg'
Learn More

Köttermann Pharmacy Fume Hood with Media

Product Code. 17302189
Click to view available options
Configuration Options:
No Ventilation Fan
Ventilation Fan
Plug Type:
GB Plug
Plug Type D (Old British Plug)
Schuko Plug
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17302189

Brand: Köttermann 45100024

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Pharmacy Fume Hood with Media is designed for the handling of hazardous substances in pharmacies and is ideally suited for working with dusty preparations or volatile chemicals.

  • For working safely with dust-forming preparations and volatile chemicals
  • Long-lasting steel construction with one-piece polyacrylic sash
  • Ergonomic features to improve workflow
  • Low, compact design for small laboratories

Specifications

Configuration Options Ventilation Fan
Dimensions (L x W x H) 640 x 900 x 1350 mm
Includes Media
Plug Type GB Plug
Voltage 230 V
Weight (Metric) 115 kg
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.