missing translation for 'onlineSavingsMsg'
Learn More

Köttermann Pharmacy Fume Hood with Media

Produktkod. 17332179
Change view
Klicka för att se tillgängliga alternativ
Configuration Options:
No Ventilation Fan
Ventilation Fan
Plug Type:
GB Plug
Plug Type D (Old British Plug)
Schuko Plug
Förpackningsstorlek:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Configuration Options Plug Type unitSize
17332179 No Ventilation Fan Plug Type D (Old British Plug) Each
17372179 No Ventilation Fan Schuko Plug Each
17312189 No Ventilation Fan GB Plug Each
17362179 Ventilation Fan Schuko Plug Each
17302189 Ventilation Fan GB Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17332179 Leverantör Köttermann Leverantörsnummer 45100013

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denna artikel kan inte returneras. Se returpolicy

This Pharmacy Fume Hood with Media is designed for the handling of hazardous substances in pharmacies and is ideally suited for working with dusty preparations or volatile chemicals.

  • For working safely with dust-forming preparations and volatile chemicals
  • Long-lasting steel construction with one-piece polyacrylic sash
  • Ergonomic features to improve workflow
  • Low, compact design for small laboratories

Specifications

Configuration Options No Ventilation Fan
Dimensions (L x W x H) 640 x 900 x 1350 mm
Includes Media
Plug Type Plug Type D (Old British Plug)
Voltage 230 V
Weight (Metric) 115 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.