missing translation for 'onlineSavingsMsg'
Learn More

Köttermann Pharmacy Fume Hood without Media

Product Code. 17392179
Change view
Click to view available options
Configuration Options:
No Ventilation Fan
Ventilation Fan
Plug Type:
GB Plug
Plug Type D (Old British Plug)
Schuko Plug
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Options Plug Type unitSize
17392179 No Ventilation Fan Schuko Plug Each
17352179 No Ventilation Fan Plug Type D (Old British Plug) Each
17332189 No Ventilation Fan GB Plug Each
17342179 Ventilation Fan Plug Type D (Old British Plug) Each
17382179 Ventilation Fan Schuko Plug Each
17322189 Ventilation Fan GB Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17392179 Supplier Köttermann Supplier No. 45100019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Pharmacy Fume Hood without Media is designed for the handling of hazardous substances in pharmacies and is ideally suited for working with dusty preparations or volatile chemicals.

  • For working safely with dust-forming preparations and volatile chemicals
  • Long-lasting steel construction with one-piece polyacrylic sash
  • Ergonomic features to improve workflow
  • Low, compact design for small laboratories

Specifications

Configuration Options No Ventilation Fan
Dimensions (L x W x H) 640 x 900 x 1350 mm
Plug Type Schuko Plug
Voltage 230 V
Weight (Metric) 115 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.