Learn More
Motic™ Phase Contrast Attachment 10X
Product Code. 15495122
Change view
Click to view available options
For Use With (Equipment):
10X Objective, B1-223ASC, B3-223PL, DMWB1-223ASC Microscopes
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15495122 10X Objective, B1-223ASC, B3-223PL, DMWB1-223ASC Microscopes Each
1 options
Product Code. 15495122 Supplier Motic™ Supplier No. 12220266
  • ONLINE EXCLUSIVE PRICE
    88.75 €
    80.41 € / Pack of 1
    Save 8.34 €
    9% Off
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Useful for improving visualization of transparent specimens via phase contrast. Motic™ Phase Contrast Attachment 10X is a simple, plug-in type attachment.
  • Improve visualization of transparent specimens.
  • Simple, plug-in type attachment.
  • For use with 10X objective.
Specifications
Type Phase Contrast Plug-in Attachment
For Use With (Equipment) 10X Objective, B1-223ASC, B3-223PL, DMWB1-223ASC Microscopes