missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Select from homogeneous and gradient polyacrylamide gels
- Separate proteins and peptides on denaturing and native
- Save time and improve reproducibility using precast gels with native or denaturing buffer strips and staining kits, buffer strips, and staining kits
- Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
- Gels are cast on a plastic backing for ease of handling
PhastGel homogenous gels
- Available in 7.5%, 12.5%, and 20% polyacrylamide with stacking gel, for flexibility in separation, extending the resolution between proteins within a narrow MW range
- A high density PhastGel (20%) is also available containing PEG to optimise separations below Mr 20000 increasing resolution
PhastGel gradient gels
- Available in concentration gradients of 4-15%,10-15% and 8-25% with stacking gel, for separation of complex mixtures, giving a good overall view of total composition within a sample
Specifications
Specifications
| Boiling Point | 125°C |
| Cassette Material | Polyester |
| Description | PhastGel Homogeneous – 12.5 |
| Gel Percentage | 12.5% |
| Gel Thickness | 0.45 mm |
| Gel Type | Tris-Acetate |
| Separation of | Protein |
| Separation Range | 230 Mr |
| Separation Type | Denaturing, Native |
| For Use With (Application) | Analytical Chemistry and Research |
| Show More |
Safety and Handling
Signal Word
- Danger||
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- H340-May cause genetic defects.
- H350-May cause cancer.
- EUH208-Contains Acrylamide. May produce an allergic reaction.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Acrylamide
- REA17-CMR1-Restricted to professional users
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?