missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PhastGel™ Buffer Strips

Product Code. 10269294 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Application):
Gel Electrophoresis
Product Type:
Buffer strip holder
PhastGel Buffer Strips-SDS
PhastGel Buffer Strips-native
Quantity:
2 Pack
20 Strips
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Product Code. For Use With (Application) Product Type Quantity unitSize
10269294 Gel Electrophoresis Buffer strip holder 2 Pack Pack of 2
11584915 Gel Electrophoresis PhastGel Buffer Strips-native 20 Strips Pack of 10
11798448 Gel Electrophoresis PhastGel Buffer Strips-SDS 20 Strips Pack of 20
3 options
This item is not returnable. View return policy

Product Code. 10269294

Brand: Cytiva 18166801

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pre-prepared 3% agarose blocks containing buffer, for use in conjunction with PhastGel gradient and homogeneous gel media to generate a multiphasic buffer system during electrophoresis

  • Ready made buffer strips eliminate the need to prepare electrophoresis buffers
  • Supplied as 10X pairs - for both cathode and anode
  • Choice of Native or Denaturing (SDS) buffer systems

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gel Electrophoresis
Quantity 2 Pack
Product Type Buffer strip holder
For Use With (Equipment) PhastSystem Automated Electrophoresis Unit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.