missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Freedom and versatility of a homemade gel with the convenience, reproducibility, increased safety, and speed of a precast gel.
- The gel is easily rehydrated in a rehydration cassette.
- Save time and improve reproducibility with precast gels, buffer strips, and staining kits.
Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits. - Cast, washed, and dried.
- Contains no residual acrylamide monomers or other contaminants from the polymerization process.
- Before use, the gel must be reswollen with a solution containing carrier ampholytes and other additives required for the intended application.
Specifications
Specifications
| Cassette Material | Polyester |
| Description | PhastGel – 10-50 |
| For Use With (Equipment) | PhastGel Cassette, Rehydration Cassette |
| Gel Percentage | 2 to 6% |
| Gel Thickness | 0.45 mm |
| Gel Type | Pharmalyte |
| Product Type | IEF Gel |
| Separation of | Protein |
| Separation Range | 10 to 50 Mr |
| Separation Type | Denaturing, Native |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?