missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Phen Green SK diacetate
GREENER_CHOICE

Product Code. 18775915
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18775915

Brand: Tocris Bioscience™ 7873/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Heavy metal ion indicator, membrane permeable

Phen Green SK diacetate is a fluorescent heavy metal ion indicator that can be used to detect various metal ions including Cu2+, Cu+, Fe2+, Hg2+, Pb2+, Cd2+, Zn2+ and Ni2+. Excitation and emission maxima (λ) are 507 and 532 nm, respectively. Intracellular Phen Green SK diacetate fluorescence is quenched upon reaction with metal ions. This product is cell permeable.

It is recommended to prepare stock solutions in DMSO.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3 ',6 '-Bis(acetyloxy)-2 ',7 '-dichloro-3-oxo-N-1,10-phenanthrolin-5-yl-spiro[isobenzofuran-1(3 H),9 '-[9 H]xanthene]-5-carboxamide
CAS 234075-45-7
Quantity 1 mg
Target Fluorescent Cell Indicators and Sensors
Molecular Formula C37H21Cl2N3O8
Purity 0.9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.