missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene, 97%

Product Code. p-180791 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
500g
50g
5g
This item is not returnable. View return policy

Product Code. 10745241

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85-01-8 | C14H10 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-01-8
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001168
InChI Key YNPNZTXNASCQKK-UHFFFAOYSA-N
Synonym phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
PubChem CID 995
ChEBI CHEBI:28851
IUPAC Name phenanthrene
SMILES C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Specifications

Melting Point 95°C to 101°C
Color White to Gray
Density 1.0630g/mL
Boiling Point 336°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05, 667
Merck Index 15, 7326
Specific Gravity 1.063
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetic acid,soluble in alcohol,ether,benzene and carbon,disulfide
Formula Weight 178.23
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Phenanthrene, 98+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.