missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene, tech. 90%

Product Code. 11424197
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424197 2500 g 2500g
11404197 100 g 100g
11414197 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424197 Supplier Thermo Scientific Alfa Aesar Supplier No. A19646.0E
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 85-01-8 | C14H10 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Phenanthrene is generally used for dye production, synthetic resin, preservative. It is also used in agrochemical and pharmaceutical intermediates.

Solubility
Soluble in alcohol, benzene, toluene, and glacial acetic acid

Notes
Light sensitive. Handle and store under inert gas. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-01-8
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001168
InChI Key YNPNZTXNASCQKK-UHFFFAOYSA-N
Synonym phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
PubChem CID 995
ChEBI CHEBI:28851
IUPAC Name phenanthrene
SMILES C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Specifications

Melting Point 97°C to 101°C
Density 0.98
Boiling Point 340°C
Flash Point 171°C (339°F)
Quantity 2500 g
UN Number UN3077
Beilstein 1905428
Merck Index 14,7212
Solubility Information Soluble in alcohol, benzene, toluene, and glacial acetic acid
Formula Weight 178.23
Percent Purity 90%
Grade Technical
Chemical Name or Material Phenanthrene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.