missing translation for 'onlineSavingsMsg'
Learn More

Phenanthridine, 98%

Product Code. 10194251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194251 5 g Glass bottle 5g
10421792 1 g Glass bottle 1g
10463785 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10194251 Supplier Thermo Scientific Acros Supplier No. 169250050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 229-87-8 | C13H9N | 179.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 229-87-8
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.22
MDL Number MFCD00004989
InChI Key RDOWQLZANAYVLL-UHFFFAOYSA-N
Synonym 6-phenanthridine, benzo c quinoline, 3,4-benzoquinoline, 9-azaphenanthrene, 3,4-benzoisoquinoline, 5-azaphenanthrene, unii-62qgs7cps6, ccris 1234, 62qgs7cps6
PubChem CID 9189
ChEBI CHEBI:36421
IUPAC Name phenanthridine
SMILES C1=CC=C2C(=C1)C=NC3=CC=CC=C23

Specifications

Melting Point 104.0°C to 108.0°C
Color White to Beige
Density 100g/mL
Boiling Point 349.0°C (769.0 mmHg)
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 20, 466
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene, dmso
Formula Weight 179.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phenanthridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenanthridine
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.