missing translation for 'onlineSavingsMsg'
Learn More

Phenazine, 98%

Product Code. 10417140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10417140 10 g Glass bottle 10g
10163130 5 g Glass bottle 5g
15199173 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10417140 Supplier Thermo Scientific Acros Supplier No. 130150100
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-82-0 | C12H8N2 | 180.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-82-0
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00005023
InChI Key PCNDJXKNXGMECE-UHFFFAOYSA-N
Synonym dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
PubChem CID 4757
ChEBI CHEBI:36674
IUPAC Name phenazine
SMILES C1=CC=C2C(=C1)N=C3C=CC=CC3=N2

Specifications

Melting Point 174.0°C to 177.0°C
Color Yellow to Brown
Boiling Point >360.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 23,223
Merck Index 15,7331
Solubility Information Solubility in water: insoluble. Other solubilities: partially soluble in alcohol and ether, moderately soluble in benzene
Formula Weight 180.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phenazine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.