missing translation for 'onlineSavingsMsg'
Learn More

Phenazine, 99+%

Product Code. 11499180
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499180 100 g 100g
11479180 5 g 5g
11489180 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499180 Supplier Thermo Scientific Alfa Aesar Supplier No. A15770.22
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-82-0 | C12H8N2 | 180.21 g/mol

Phenazine is the parent substance of many dyestuffs, such as the toluylene red, indulines and safranines. It can be applied in therapeutic uses, used as antipsychotic and antiemetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-82-0
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00005023
InChI Key PCNDJXKNXGMECE-UHFFFAOYSA-N
Synonym dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
PubChem CID 4757
ChEBI CHEBI:36674
IUPAC Name phenazine
SMILES C1=CC=C2C(=C1)N=C3C=CC=CC3=N2

Specifications

Melting Point 175°C to 177°C
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 180.21
Percent Purity ≥99%
Physical Form Crystals or Powder or Crystalline Powder or Needles
Chemical Name or Material Phenazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.