missing translation for 'onlineSavingsMsg'
Learn More

Phenazine ethosulfate, ∽95%, MP Biomedicals™

Product Code. 11465955 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465955 5 g 5g
11455955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465955 Supplier MP Biomedicals Supplier No. 0210095605
  • ONLINE EXCLUSIVE PRICE
    479.00 €
    453.60 € / 5g
    Save 25.40 €
    5% Off
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10510-77-7
Molecular Formula C16H18N2O4S
Molecular Weight (g/mol) 334.39
MDL Number MFCD00050286
InChI Key VDJKJPMLWJWQIH-UHFFFAOYSA-M
Synonym phenazine ethosulfate, 5-ethylphenazine, n-ethylphenazonium ethosulfate, 5-ethylphenazinium ethyl sulphate, 5-ethylphenazin-5-ium ethyl sulfate, 5-ethylphenazinium ethylsulfate, phenazinium, 5-ethyl-, ethyl sulfate, 5-ethylphenazine, ethyl sulfate salt, ccris 5619, n-ethyldibenzopyrazine ethyl sulfate salt
PubChem CID 82688
IUPAC Name 5-ethylphenazin-5-ium;ethyl sulfate
SMILES CC[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.CCOS(=O)(=O)[O-]

Specifications

Color Orange-Yellow
Assay Percent Range ∽ 95%
Quantity 5 g
Formula Weight 334.4
Physical Form Powder
Chemical Name or Material Phenazine ethosulfate
compliance-icons
Product Identifier
  • Phenazine Ethosulfate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Phenazine Ethosulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.