missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl Isothiocyanate, TRC

Product Code. 30138150
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138150 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30138150 Supplier Toronto Research Chemicals Supplier No. TRCP29610025G
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2257-09-2
Molecular Formula C9 H9 N S
Quantity 25 g
Synonym Benzene, (2-isothiocyanatoethyl)-, Isothiocyanic acid, phenethyl ester (6CI,7CI,8CI), (2-Isothiocyanatoethyl)benzene, 1-(2-Isothiocyanatoethyl)benzene, 2-Phenylethyl isothiocyanate, Beta-phenylethyl isothiocyanate, JC 5411, NSC 87868, Phenethyl isothiocyanate, Phenethyl mustard oil, Phenylethyl isothiocyanate, Phenylethyl mustard oil, β-Phenethyl isothiocyanate, β-Phenylethyl isothiocyanate
InChI Formula InChI=1S/C9H9NS/c11-8-10-7-6-9-4-2-1-3-5-9/h1-5H,6-7H2
SMILES S=C=NCCc1ccccc1
IUPAC Name 2-isothiocyanatoethylbenzene
Molecular Weight (g/mol) 163.24
Formula Weight 163.0456
Chemical Name or Material Phenylethyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.