missing translation for 'onlineSavingsMsg'
Learn More

Phenethyleisothiocyanate, 97%, Thermo Scientific™

Product Code. 10608783
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10608783 10 g Amber Glass Bottle 10g
10216004 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10608783 Supplier Thermo Scientific Maybridge Supplier No. TL00242EA
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2257-09-2
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00004821
InChI Key IZJDOKYDEWTZSO-UHFFFAOYSA-N
Synonym 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
PubChem CID 16741
ChEBI CHEBI:351346
IUPAC Name 2-isothiocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=S

Specifications

CAS Min % 97.0
Density 1.1000g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C6H5CH2CH2NCS
Quantity 10 g
Merck Index 15, 7340
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material Phenethyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.