missing translation for 'onlineSavingsMsg'
Learn More

Phenidone, TRC

Product Code. 30411560
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411560 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411560 Supplier Toronto Research Chemicals Supplier No. TRCP2972535G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenidone
CAS 92-43-3
Molecular Formula C9H10N2O
Quantity 5 g
Synonym 1-Phenyl-3-pyrazolidinone, 1-Phenyl-3-oxopyrazolidine, 1-Phenyl-3-pyrazolidone, 1-Phenylpyrazolidine-3-one, Fenidon, Phenidone
InChI Formula InChI=1S/C9H10N2O/c12-9-6-7-11(10-9)8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,12)
SMILES O=C1CCN(N1)c2ccccc2
IUPAC Name 1-phenylpyrazolidin-3-one
Molecular Weight (g/mol) 162.19
Formula Weight 162.0793
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.