missing translation for 'onlineSavingsMsg'
Learn More

Phenol-d1, 95% (Isotopic), Thermo Scientific™

Product Code. 15447987
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447987 5 g 5g
15437987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15447987 Supplier Thermo Scientific Alfa Aesar Supplier No. H51100.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-66-3
Molecular Formula C6H5DO
Molecular Weight (g/mol) 95.12
MDL Number MFCD01075468
InChI Key ISWSIDIOOBJBQZ-DYCDLGHISA-N
Synonym phenol-d, phenol-d1, o-2h phenol, phenol-od, 2 h oxy benzene
PubChem CID 12205409
IUPAC Name deuteriooxybenzene
SMILES [2H]OC1=CC=CC=C1

Specifications

Density 1.082
Sensitivity Air and Light
Quantity 5 g
Melting Point 40°C to 42°C
Boiling Point 182°C
UN Number UN1671
Formula Weight 95.12
Percent Purity 95%
Chemical Name or Material Phenol-d1, Isotopic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.