missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red, ACS reagent

Product Code. 10035950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10035950 5 g Glass bottle 5g
10157750 10 g Glass bottle 10g
10299473 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10035950 Supplier Thermo Scientific Chemicals Supplier No. 417240050
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 143-74-8 | C19H14O5S | 354.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonephthalein
PubChem CID 4766
ChEBI CHEBI:31991
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point >300.0°C
Clarity passes test
Beilstein 19, 91
Merck Index 15, 7360
Solubility Information Solubility in water: 0.77g/L. Other solubilities: soluble in ethanol
Infrared Spectrum Authentic
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Formula Weight 354.38
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 6.8 (yellow) to pH 8.2 (red)
Color Colorless to Yellow
Quantity 5 g
Physical Form Liquid After Melting
Chemical Name or Material Phenol Red, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Phenol Red
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.