missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, 0.5% (w/v), Ricca Chemical

Product Code. 31749488
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31749488 500 mL Natural Poly Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31749488 Supplier Ricca Chemical Company Supplier No. R5724500500A
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 34487-61-1, 7732-18-5
CAS Min % 98.0, 0.49
CAS Max % 100.0, 0.51
Packaging Natural Poly Bottle
Molecular Formula C19H13NaO5S
Color Orange/Red
pH 6 to 7.3
Quantity 500 mL
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
Physical Form Liquid
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
Molecular Weight (g/mol) Mixture
PubChem CID 23686673
Percent Purity 0.5% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 0.5% (w/v) Aqueous Solution
Chemical Name or Material Phenol Red Indicator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.