missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, ACS Reagent, Solstice

Product Code. 15643830
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15643830 100 g Glass Bottle 100g
15653830 10 g Glass Bottle 10g
15663830 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15643830 Supplier Honeywell-Fluka Supplier No. 32661100G
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Indicator, ACS reagent

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonphthalein
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Packaging Glass Bottle
Visual Transition Interval 6.8 to 8.2 (yellow to red)
UN Number NONH for all modes of transport
Beilstein 326470
Color Red
Quantity 100 g
Physical Form Powder
Formula Weight 354.38g/mol
Grade ACS Reagent
Chemical Name or Material Phenol Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.