missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red, sodium salt, pure, water soluble, indicator

Product Code. 10279223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10279223 5 g Glass bottle 5g
10418120 25 g Glass bottle 25g
11986801 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Product Code. 10279223 Supplier Thermo Scientific Chemicals Supplier No. 151440050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Melting Point 285.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Extinction 1600 (558nm, in buffer) min.
Visual Transition Interval pH 6.8 (yellow) to pH 8.2 (red)
Color Brown to Red
Solubility Information Solubility in water: soluble.
Quantity 5 g
Physical Form Amorphous Powder
Formula Weight 376.35
Grade Pure
Chemical Name or Material Phenol Red, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Phenol Red, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.