missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Phenolphthalein

Product Code. 16422419
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422419 Supplier MedChemExpress Supplier No. HYD0211
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Phenolphthalein is a widely applied but toxic indicator dye.

Specifications

Quantity 1 g
Storage Requirements 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Solubility DMSO : 100 mg/mL (314.15 mM; Need ultrasonic and warming)
Shipping Condition Room temperature
CAS 77-09-8
Molecular Weight (g/mol) 318.32 g/mol
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.