missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenolphthalein, 98%

Product Code. 11488996
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488996 1000 g 1000g
11498996 50 g 50g
11409006 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488996 Supplier Thermo Scientific Chemicals Supplier No. A17135.0B
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-09-8 | C20H14O4 | 318.33 g/mol

Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extent of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extent of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

Solubility
Soluble in alcohol. Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.

Notes
Acid-base indicator: pH 8.0 - 10.0.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
PubChem CID 4764
ChEBI CHEBI:34914
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 258°C to 262°C
Density 1.299
Quantity 1000 g
Beilstein 284423
Merck Index 14,7243
Solubility Information Soluble in alcohol. Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Formula Weight 318.33
Percent Purity 98%
Chemical Name or Material Phenolphthalein
compliance-icons
Product Identifier
  • Phenolphthalein
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Phenolphthalein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.