missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein, ACS reagent, Thermo Scientific Chemicals

Product Code. 10667211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10667211 100 g Plastic bottle 100g
10289840 500 g Plastic bottle 500g
10135513 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10667211 Supplier Thermo Scientific Acros Supplier No. 417181000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-09-8 | C20H14O4 | 318.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 258.0°C to 263.0°C
Clarity Passes Test
Infrared Spectrum Authentic
Packaging Plastic bottle
Vapor Pressure Vapor Pressure: Negligible
Visual Transition Interval from pH 8.0 (colorless) to pH 10 (red)
Beilstein 18, 143
Density 1.299g/mL
Merck Index 15, 7356
Color White
Solubility Information Solubility in water: <0.1%. Other solubilities: 1g/12mL alcohol, 1g/100mL ether, soluble in alkalies, soluble in chloroform
Quantity 100 g
Physical Form Crystalline Powder
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Formula Weight 318.32
Grade ACS Reagent
Chemical Name or Material Phenolphthalein
Show More Show Less
compliance-icons
Product Identifier
  • Phenolphthalein
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.