missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Indicator, 1% (w/v) in Methanol, Neutralized, Ricca Chemical

Product Code. 31940494
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31940494 1 L Natural Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 31940494 Supplier Ricca Chemical Company Supplier No. R56210001A
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 77-09-8 , 67-56-1 , 7732-18-5
Molecular Formula C20H14O4
Molecular Weight (g/mol) Mixture
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

CAS Min % 97.0, 0.5
CAS Max % 100.0, 1.5
Packaging Natural Poly Bottle
Color Colorless to Pink
pH 8
Quantity 1 L
Physical Form Liquid
Percent Purity 1% W/V
Grade Indicator
Chemical Name or Material Phenolphthalein Indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.