missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein, TRC

Product Code. 30136328
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136328 250 g 250g
30132833 25 g 25g
30007538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136328 Supplier Toronto Research Chemicals Supplier No. TRCP318010250G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20 H14 O4
Molecular Weight (g/mol) 318.32
Synonym Phenolphthalein, 3,3-bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one, 1(3H)-Isobenzofuranone, 3,3-bis(4-hydroxyphenyl)-, Phenolphthalein (8CI), 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranone, 3,3-Bis(4-hydroxyphenyl)phthalide, 3,3-Bis(p-hydroxyphenyl)phthalide, Euchessina, Koprol, Laxogen, Lilo, NSC 10464, NSC 215214, Phthalimetten, Phthalin, Purga, Purgen, Purgophen, Spulmako-lax, TY 9, Trilax
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
SMILES Oc1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc(O)cc4

Specifications

Quantity 250 g
InChI Formula InChI=1S/C20H14O4/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20/h1-12,21-22H
Formula Weight 318.0892
Percent Purity >95%
Chemical Name or Material Phenolphthalein
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.