missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein, TRC

Product Code. 30007538
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007538 5 g 5g
30136328 250 g 250g
30132833 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007538 Supplier Toronto Research Chemicals Supplier No. TRCP3180105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Formula Weight 318.0892
InChI Formula InChI=1S/C20H14O4/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20/h1-12,21-22H
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Molecular Formula C20 H14 O4
Molecular Weight (g/mol) 318.32
Percent Purity >95%
SMILES Oc1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc(O)cc4
Synonym Phenolphthalein, 3,3-bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one, 1(3H)-Isobenzofuranone, 3,3-bis(4-hydroxyphenyl)-, Phenolphthalein (8CI), 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranone, 3,3-Bis(4-hydroxyphenyl)phthalide, 3,3-Bis(p-hydroxyphenyl)phthalide, Euchessina, Koprol, Laxogen, Lilo, NSC 10464, NSC 215214, Phthalimetten, Phthalin, Purga, Purgen, Purgophen, Spulmako-lax, TY 9, Trilax
Chemical Name or Material Phenolphthalein
Grade HPLC
Show More Show Less

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.