missing translation for 'onlineSavingsMsg'
Learn More

Phenosafranin

Product Code. 10022021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10022021 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10022021 Supplier Thermo Scientific Acros Supplier No. 339340050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81-93-6 | C18H15ClN4 | 322.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

CAS 81-93-6
Melting Point >300.0°C
Color Green
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C18H15ClN4
MDL Number MFCD00036335
Quantity 5 g
Merck Index 15, 7363
Synonym phenosafranin, phenosafranine, phenosafranine, chloride, phenosafrinine, phenosofranine, phenosofrinine, safranin b extra, safrinin b extra, 3,7-diamino-5-phenylphenazinium chloride, schultz no. 958
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
InChI Key SOUHUMACVWVDME-UHFFFAOYSA-N
SMILES [Cl-].NC1=CC2=[N+](C3=CC=CC=C3)C3=CC(N)=CC=C3N=C2C=C1
IUPAC Name 10-phenylphenazin-10-ium-2,8-diamine;chloride
Molecular Weight (g/mol) 322.80
PubChem CID 65732
ChEBI CHEBI:33601
Formula Weight 322.8
Physical Form Crystalline Powder
Chemical Name or Material Phenosafranin
Show More Show Less
compliance-icons
Product Identifier
  • Phenosafranin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.