missing translation for 'onlineSavingsMsg'
Learn More

Phenothiazine, 99%

Product Code. 10308613 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10308613 5 kg Plastic drum 5kg
10214800 500 g Plastic Bottle 500g
10695942 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10308613 Supplier Thermo Scientific Acros Supplier No. 130210050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-84-2 | C12H9NS | 199.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-84-2
Molecular Formula C12H9NS
Molecular Weight (g/mol) 199.28
MDL Number MFCD00005015
InChI Key WJFKNYWRSNBZNX-UHFFFAOYSA-N
Synonym phenothiazine, thiodiphenylamine, feeno, dibenzothiazine, phenosan, phenthiazine, dibenzo-1,4-thiazine, penthazine, souframine, agrazine
PubChem CID 7108
ChEBI CHEBI:37931
IUPAC Name 10H-phenothiazine
SMILES C1=CC=C2C(=C1)NC3=CC=CC=C3S2

Specifications

Melting Point 185.0°C
Color Yellow
Boiling Point 371.0°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic drum
Quantity 5 kg
Beilstein 27, 63
Merck Index 15, 7364
Solubility Information Solubility in water: 2mg/l (25°C). Other solubilities: freely soluble in hot acetic acid, freely soluble in benzene and ether, slightly soluble in alcohol and mineral oils, practically insoluble in chloroform and petroleum, ether
Formula Weight 199.28
Percent Purity 99%
Physical Form Prills or Beads
Chemical Name or Material Phenothiazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenothiazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.