missing translation for 'onlineSavingsMsg'
Learn More

Phenprobamate, TRC

Product Code. 30422406
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422406 5 g -
30410642 10 g -
30417565 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422406 Supplier Toronto Research Chemicals Supplier No. TRCP3188585G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-31-4
Molecular Formula C10 H13 N O2
Molecular Weight (g/mol) 179.22
Synonym Benzenepropanol, 1-carbamate, 1-Propanol, 3-phenyl-, carbamate (8CI), Benzenepropanol, carbamate (9CI), Carbamic acid, 3-phenylpropyl ester (6CI,7CI), 3-Phenyl-1-propanol carbamate, 3-Phenylpropyl carbamate, Ansepron, Eirenal, Extacol, Gamaquil, MH 532, NSC 44682, NSC 50538, NSC 64270, Palmita, Phenprobamate, Proformiphen, Quamaquil, Spantol, Tranquil, γ-Phenylpropyl carbamate
IUPAC Name 3-phenylpropyl carbamate
SMILES NC(=O)OCCCc1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H13NO2/c11-10(12)13-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H2,11,12)
Formula Weight 179.0946
Chemical Name or Material Phenprobamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.