missing translation for 'onlineSavingsMsg'
Learn More

Phensuximide, TRC

Product Code. 17990935
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17990935 50 mg 50mg
17930605 25 mg 25mg
17952175 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17990935 Supplier Toronto Research Chemicals Supplier No. TRCP31890050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-34-0
Molecular Formula C11 H11 N O2
Molecular Weight (g/mol) 189.21
Synonym 2,5-Pyrrolidinedione, 1-methyl-3-phenyl-, Succinimide, N-methyl-2-phenyl- (6CI,7CI,8CI), (±)-Phensuximide, 1-Methyl-3-phenyl-2,5-pyrrolidinedione, 1-Methyl-3-phenylsuccinimide, Epimid, Lifene, Milontin, Milonton, Mirontin, N-Methyl-2-phenylsuccinimide, N-Methyl-3-phenylpyrrolidinedione, N-Methyl-3-phenylsuccinimide, N-Methyl-α-phenylsuccinimide, PM 334, Phensuximid, Phensuximide, RS-Phensuximide, Racemic phensuximide, Succitimal
IUPAC Name 1-methyl-3-phenylpyrrolidine-2,5-dione
SMILES CN1C(=O)CC(C1=O)c2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H11NO2/c1-12-10(13)7-9(11(12)14)8-5-3-2-4-6-8/h2-6,9H,7H2,1H3
Formula Weight 189.079
Chemical Name or Material Phensuximide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.