missing translation for 'onlineSavingsMsg'
Learn More

Phentolamine Methanesulfonate Salt, TRC

Product Code. 30423431
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423431 250 mg 250mg
30411724 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30423431 Supplier Toronto Research Chemicals Supplier No. TRCP302560250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-28-1
Molecular Formula C17 H19 N3 O . C H4 O3 S
Molecular Weight (g/mol) 377.46
Synonym Phentolamine mesilate, 3-[[(4,5-Dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]phenol methanesulfonate, Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, methanesulfonate (1:1), Phenol, 3-[[(4,5-dihydro-1H-imidazol-2-yl)methyl](4-methylphenyl)amino]-, monomethanesulfonate (salt) (9CI), Phenol, m-[N-(2-imidazolin-2-ylmethyl)-p-toluidino]-, monomethanesulfonate (salt) (8CI), Phentolamine mesylate, Phentolamine methanesulfonate, Phentolamine methanesulphonate, Regitin methanesulphonate, Regitine mesylate, Regitine methanesulfonate, Rogitine, Vasomax, m-[N-(2-Imidazolin-2-ylmethyl)-p-toluidino]phenol methanesulfonate
IUPAC Name 3-[N-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-methylanilino]phenol;methanesulfonic acid
SMILES Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3.CS(=O)(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H19N3O.CH4O3S/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;1-5(2,3)4/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H3,(H,2,3,4)
Formula Weight 377.1409
Chemical Name or Material Phentolamine Mesilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.