missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals

Product Code. 15410567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410567 1 g 1g
15420567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410567 Supplier Thermo Scientific Alfa Aesar Supplier No. J66355.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23661-28-1 | Molecular Formula: C20H24O9S | Molecular Weight: 440.463 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23661-28-1
Molecular Formula C20H24O9S
Molecular Weight (g/mol) 440.463
MDL Number MFCD00135159
InChI Key JCKOUAWEMPKIAT-OBKDMQGPSA-N
Synonym 2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenyl-1-thio-, a-d-glucopyranoside tetraacetate, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate, 1-phenyl-1-thio-beta-d-glucopyranoside tetraacetate
PubChem CID 6364823
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 116°C to 119°C
Color White
Quantity 1 g
Formula Weight 440.47
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.