missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2-bromoacetate, 97%, BLD Pharm™

Product Code. 31111677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31111677 25 g
31093055 100 g
31098424 5 g
31102669 10 g
31102929 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31111677 Supplier BLD Pharm Supplier No. BD14755725g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 620-72-4
Molecular Formula C8H7BrO2
MDL Number MFCD00192391
Quantity 25 g
SMILES O=C(OC1=CC=CC=C1)CBr
Molecular Weight (g/mol) 215.04
Percent Purity 97%
Chemical Name or Material Phenyl 2-bromoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.