missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2-pyridyl ketoxime, 98%, Thermo Scientific™

Product Code. 10157962 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10157962 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10157962 Supplier Thermo Scientific Acros Supplier No. 373440050
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1826-28-4
Melting Point 154.0°C to 156.0°C
Color Pink
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C12H10N2O
MDL Number MFCD00006310
Quantity 5 g
Synonym phenylpyridylketoxime, phenyl pyridin-2-yl methanone oxime, phenyl 2-pyridyl ketone oxime, methanone, phenyl-2-pyridinyl-, oxime, 2-benzoylpyridiine ketoxime, phenyl 2-pyridyl methanone oxime, 2-benzoylpyridine oxime, e-phenyl-2-pyridyl ketone oxime, hydroxyimino phenyl-2-pyridylmethane, ketone, phenyl 2-pyridyl, oxime 8ci
InChI Key WBSIGZZXFQOEJK-QXMHVHEDSA-N
SMILES O=N\C(=C1/NC=CC=C1)C1=CC=CC=C1
IUPAC Name (2E)-2-[nitroso(phenyl)methylidene]-1H-pyridine
Molecular Weight (g/mol) 198.23
PubChem CID 5397197
Formula Weight 198.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 2-pyridyl ketoxime
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.