missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Chloroformate, TRC
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Chemical Name or Material | Phenyl Chloroformate |
| CAS | 1885-14-9 |
| Molecular Formula | C7H5ClO2 |
| Quantity | 250 g |
| Synonym | Carbonochloridic Acid Phenyl Ester, NSC 210946, Phenyl Carbonochloridate, Phenyl Chlorocarbonate, Phenyl Chloroformate, Phenylchloroformic Acid, Phenyloxycarbonyl Chloride |
| InChI Formula | InChI=1S/C7H5ClO2/c8-7(9)10-6-4-2-1-3-5-6/h1-5H |
| SMILES | ClC(=O)Oc1ccccc1 |
| IUPAC Name | phenyl carbonochloridate |
| Molecular Weight (g/mol) | 156.57 |
| Formula Weight | 155.9978 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction