missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Chloroformate, TRC

Product Code. 30417419
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30417419 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30417419

Brand: Toronto Research Chemicals TRCP31988050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenyl Chloroformate
CAS 1885-14-9
Molecular Formula C7H5ClO2
Quantity 50 g
Synonym Carbonochloridic Acid Phenyl Ester, NSC 210946, Phenyl Carbonochloridate, Phenyl Chlorocarbonate, Phenyl Chloroformate, Phenylchloroformic Acid, Phenyloxycarbonyl Chloride
InChI Formula InChI=1S/C7H5ClO2/c8-7(9)10-6-4-2-1-3-5-6/h1-5H
SMILES ClC(=O)Oc1ccccc1
IUPAC Name phenyl carbonochloridate
Molecular Weight (g/mol) 156.57
Formula Weight 155.9978
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.